Please login to your online account to display your discounted pricing

anti-DYRK3, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DYRK3 (within residues 500-600). [Swiss-Prot No. O43781]
Antigen DYRK3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11094876

 
novus biologicals canada
NB11094876
Unlabeled 0.1mL Each for $309.05

NB11094876T

 
novus biologicals canada
NB11094876SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DYRK3 (within residues 500-600). [Swiss-Prot No. O43781]
Antigen DYRK3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human and monkey. Does not react with mouse and rat. Immunogen sequence has 94% homology to bovine.
Gene Accession No. O43781
Gene ID (Entrez) 8444
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL