Please login to your online account to display your discounted pricing

anti-DYRK3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11094876SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11094876T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8444
Formulation Tris-citrate/phosphate, pH 7-8
Primary or Secondary Primary
Conjugate Unlabeled
Gene Accession No. O43781
Concentration 1mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.025mL
Test Specificity Human and monkey. Does not react with mouse and rat. Immunogen sequence has 94% homology to bovine.
Immunogen Synthetic peptide made to an internal portion of human DYRK3 (within residues 500-600). [Swiss-Prot No. O43781]
Host Species Rabbit
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Primate
Antigen DYRK3
Regulatory Status RUO