Please login to your online account to display your discounted pricing

anti-DYRK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11094452

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11094452

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8445
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Formulation Tris-citrate/phosphate, pH 7-8
Test Specificity Human and chicken. Does not react with mouse and rat. Immunogen sequence has 91% homology to Zebrafish.
Concentration 1mg/mL
Gene Accession No. Q92630
Conjugate Unlabeled
Host Species Rabbit
Antigen DYRK2
Cross Reactivity Chicken
Cross Reactivity Human
Quantity 0.1mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of human DYRK2 (within residues 400-500). [Swiss-Prot No. Q92630]