Please login to your online account to display your discounted pricing

anti-DYRK2, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DYRK2 (within residues 400-500). [Swiss-Prot No. Q92630]
Antigen DYRK2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11094452T

 
novus biologicals canada
NB11094452SS
Unlabeled 0.025mL Each for $109.00

NB11094452

 
novus biologicals canada
NB11094452
Unlabeled 0.1mL Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DYRK2 (within residues 400-500). [Swiss-Prot No. Q92630]
Antigen DYRK2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human and chicken. Does not react with mouse and rat. Immunogen sequence has 91% homology to Zebrafish.
Gene Accession No. Q92630
Gene ID (Entrez) 8445
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL