Please login to your online account to display your discounted pricing

anti-DYRK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11094452SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11094452T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8445
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q92630
Concentration 1mg/mL
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Conjugate Unlabeled
Test Specificity Human and chicken. Does not react with mouse and rat. Immunogen sequence has 91% homology to Zebrafish.
Cross Reactivity Chicken
Cross Reactivity Human
Regulatory Status RUO
Antigen DYRK2
Quantity 0.025mL
Immunogen Synthetic peptide made to an internal portion of human DYRK2 (within residues 400-500). [Swiss-Prot No. Q92630]
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified