Please login to your online account to display your discounted pricing

anti-DYKDDDDK Epitope Tag, Polyclonal, Novus Biologicals™

$307.00 - $307.00

Specifications

Host Species Chicken
Immunogen Chickens were immunized with ECS enterokinase cleavage site (xxxDDDDK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen DYKDDDDK Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600343

 
novus biologicals canada
NB600343
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Chicken
Immunogen Chickens were immunized with ECS enterokinase cleavage site (xxxDDDDK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Antigen DYKDDDDK Epitope Tag
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation PBS
Concentration 1.0mg/mL