Please login to your online account to display your discounted pricing

anti-DYKDDDDK Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6003431MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6003431MG

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Concentration 1.0mg/mL
Antigen DYKDDDDK Epitope Tag
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation PBS
Primary or Secondary Primary
Conjugate Unlabeled
Immunogen Chickens were immunized with ECS enterokinase cleavage site (xxxDDDDK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Host Species Chicken
Regulatory Status RUO
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Non-species specific