Please login to your online account to display your discounted pricing

anti-DYKDDDDK Epitope Tag, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600343

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600343

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Concentration 1.0mg/mL
Format Affinity Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Cross Reactivity Non-species specific
Monoclonal or Polyclonal Polyclonal
Host Species Chicken
Quantity 0.1mg
Antigen DYKDDDDK Epitope Tag
Immunogen Chickens were immunized with ECS enterokinase cleavage site (xxxDDDDK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary