Please login to your online account to display your discounted pricing

anti-dUTPase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056624SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056624T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1854
Gene Accession No. P33316
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Conjugate Unlabeled
Host Species Rabbit
Immunogen This antibody was developed against a mixture of KLH-conjugated synthetic peptides corresponding to amino acids 25-44 and 113-130 of human dUTPase.
Applications Western Blotting
Quantity 0.05mL
Format Purified
Regulatory Status RUO
Antigen dUTPase