Please login to your online account to display your discounted pricing

anti-DUSP8, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056410SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056410T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1850
Gene Accession No. 13202
Host Species Rabbit
Test Specificity Reacts with Human. Predicted to react with Mouse and Rat.
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Cross Reactivity Human
Primary or Secondary Primary
Applications Western Blotting
Regulatory Status RUO
Format Purified
Monoclonal or Polyclonal Polyclonal
Antigen DUSP8
Immunogen A portion of amino acids 250-300 of human DUSP8 were used as the immunogen.
Quantity 0.025mg