Please login to your online account to display your discounted pricing

anti-DUSP8, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056410

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056410

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. 13202
Gene ID (Entrez) 1850
Formulation PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Format Purified
Test Specificity Reacts with Human. Predicted to react with Mouse and Rat.
Conjugate Unlabeled
Concentration 0.5mg/mL
Host Species Rabbit
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Human
Applications Western Blotting
Primary or Secondary Primary
Antigen DUSP8
Immunogen A portion of amino acids 250-300 of human DUSP8 were used as the immunogen.