Please login to your online account to display your discounted pricing

anti-DUSP8, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human DUSP8 were used as the immunogen.
Antigen DUSP8
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056410T

 
novus biologicals canada
NB10056410SS
Unlabeled 0.025mg Each for $139.00

NB10056410

 
novus biologicals canada
NB10056410
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human DUSP8 were used as the immunogen.
Antigen DUSP8
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Reacts with Human. Predicted to react with Mouse and Rat.
Gene Accession No. 13202
Gene ID (Entrez) 1850
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL