Please login to your online account to display your discounted pricing

anti-DUSP18, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056481SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056481T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 150290
Gene Accession No. Q8NEJ0
Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Concentration 0.5mg/mL
Test Specificity Reacts with Bovine, Chimpanzee, Human, Orangutan and Rhesus Monkey. 92% sequence identity with canine and 85% with rat proteins.
Immunogen A portion of amino acids 1-50 of human DUSP18 was used as the immunogen.
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.025mg
Format Purified
Regulatory Status RUO
Antigen DUSP18
Host Species Rabbit
Applications Western Blotting
Conjugate Unlabeled