Please login to your online account to display your discounted pricing

anti-DUSP18, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056481

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056481

  • / Each
 

Description & Specifications

Specifications

Antigen DUSP18
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q8NEJ0
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human DUSP18 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 150290
Test Specificity Reacts with Bovine, Chimpanzee, Human, Orangutan and Rhesus Monkey. 92% sequence identity with canine and 85% with rat proteins.
Target Species Bovine
Target Species Human
Target Species Primate