Please login to your online account to display your discounted pricing

anti-DUSP18, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human DUSP18 was used as the immunogen.
Antigen DUSP18
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056481

 
novus biologicals canada
NB10056481
Unlabeled 0.1mg Each for $255.80

NB10056481T

 
novus biologicals canada
NB10056481SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human DUSP18 was used as the immunogen.
Antigen DUSP18
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Reacts with Bovine, Chimpanzee, Human, Orangutan and Rhesus Monkey. 92% sequence identity with canine and 85% with rat proteins.
Gene Accession No. Q8NEJ0
Gene ID (Entrez) 150290
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL