missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DUSP13, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human DUSP13 was used as the immunogen.
Antigen DUSP13
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056492 Novus Biologicals Canada
NB10056492
0.1mg Each for $255.80
NB10056492T Novus Biologicals Canada
NB10056492SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
DUSP13
Purified
RUO
This will detect isoform 3 of human DUSP13. It will not detect isoforms 1 or 2. Cross reacts with Chimpanzee and Human. Predicted to react with Bovine and Rhesus Monkey.
PBS containing 0.2% gelatin
A portion of amino acids 50-100 of human DUSP13 was used as the immunogen.
Polyclonal
Primary
Western Blot
51207