Please login to your online account to display your discounted pricing

anti-DUSP13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056492SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056492T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51207
Monoclonal or Polyclonal Polyclonal
Test Specificity This will detect isoform 3 of human DUSP13. It will not detect isoforms 1 or 2. Cross reacts with Chimpanzee and Human. Predicted to react with Bovine and Rhesus Monkey.
Conjugate Unlabeled
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human DUSP13 was used as the immunogen.
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.025mg
Regulatory Status RUO
Format Purified
Antigen DUSP13