Please login to your online account to display your discounted pricing

anti-DUSP13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056492

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056492

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51207
Formulation PBS containing 0.2% gelatin
Conjugate Unlabeled
Format Purified
Test Specificity This will detect isoform 3 of human DUSP13. It will not detect isoforms 1 or 2. Cross reacts with Chimpanzee and Human. Predicted to react with Bovine and Rhesus Monkey.
Regulatory Status RUO
Quantity 0.1mg
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Primate
Primary or Secondary Primary
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human DUSP13 was used as the immunogen.
Antigen DUSP13