Please login to your online account to display your discounted pricing

anti-Dtk/TYRO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP128635

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP128635

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7301
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Human and mouse. Immunogen sequence has 94% identity to rat.
Concentration 1.29mg/mL
Gene Accession No. Q06418
Monoclonal or Polyclonal Polyclonal
Antigen Dtk/TYRO3
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot No. Q06418]
Quantity 0.1mL
Format Affinity Purified
Regulatory Status RUO