Please login to your online account to display your discounted pricing

anti-Dtk/TYRO3, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 470-520 of human TYRO3 was used as the immunogen.
Antigen Dtk/TYRO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224520

 
novus biologicals canada
NBP224520
Unlabeled 0.1mg Each for $257.41

NBP224520SS

 
novus biologicals canada
NBP224520SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 470-520 of human TYRO3 was used as the immunogen.
Antigen Dtk/TYRO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat, dog, monkey orangutan, bovine and S. cerevisiae, and 93% in mouse and Xenopus.
Gene Accession No. Q06418
Gene ID (Entrez) 7301
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL