Please login to your online account to display your discounted pricing

anti-Dtk/TYRO3, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot No. Q06418]
Antigen Dtk/TYRO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP128635

 
novus biologicals canada
NBP128635
Unlabeled 0.1mL Each for $329.00

NBP128635SS

 
novus biologicals canada
NBP128635SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human TYRO3 protein (within residues 100-250). [Swiss-Prot No. Q06418]
Antigen Dtk/TYRO3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human and mouse. Immunogen sequence has 94% identity to rat.
Gene Accession No. Q06418
Gene ID (Entrez) 7301
Formulation PBS, 30% glycerol
Concentration 1.29mg/mL