Please login to your online account to display your discounted pricing

anti-Dtk/TYRO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224520SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224520SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7301
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL
Gene Accession No. Q06418
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat, dog, monkey orangutan, bovine and S. cerevisiae, and 93% in mouse and Xenopus.
Regulatory Status RUO
Conjugate Unlabeled
Quantity 0.025mg
Antigen Dtk/TYRO3
Format Affinity Purified
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Western Blotting
Immunogen A portion of amino acids 470-520 of human TYRO3 was used as the immunogen.
Primary or Secondary Primary