Please login to your online account to display your discounted pricing

anti-Dtk/TYRO3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224520

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224520

  • / Each
 

Description & Specifications

Specifications

Antigen Dtk/TYRO3
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q06418
Host Species Rabbit
Immunogen A portion of amino acids 470-520 of human TYRO3 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7301
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat, dog, monkey orangutan, bovine and S. cerevisiae, and 93% in mouse and Xenopus.
Target Species Human
Target Species Murine
Target Species Rat