Please login to your online account to display your discounted pricing

anti-DRP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11055237SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11055237T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10059
Format Affinity Purified
Gene Accession No. O00429
Primary or Secondary Primary
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 23658809). 100% sequence identity with monkey and hamster.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen DRP1
Conjugate Unlabeled
Concentration 1mg/mL
Formulation Tris-glycine, 150mM NaCl
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide made to an N-terminal region within residues 1-100 of the human DRP1 protein. [Swiss-Prot No. O00429]