Please login to your online account to display your discounted pricing

anti-DRP1, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region within residues 1-100 of the human DRP1 protein. [Swiss-Prot No. O00429]
Antigen DRP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11055237T

 
novus biologicals canada
NB11055237SS
Unlabeled 0.025mL Each for $139.00

NB11055237

 
novus biologicals canada
NB11055237
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region within residues 1-100 of the human DRP1 protein. [Swiss-Prot No. O00429]
Antigen DRP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 23658809). 100% sequence identity with monkey and hamster.
Gene Accession No. O00429
Gene ID (Entrez) 10059
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL