Please login to your online account to display your discounted pricing

DRAM Polyclonal Antibody, Invitrogen™


DRAM Antibody, OSD00006W, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSD00006W

Catalog No. PIOSD00006W

  • / Each
 

Description & Specifications

Specifications

Antigen DRAM
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Formulation 150μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q8N682, Q9DC58
Gene Alias Damage-regulated autophagy modulator, FLJ11259
Gene Symbols DRAM, DRAM1
Host Species Rabbit
Immunogen A synthetic peptide from the internal region of human DRAM
Quantity 150μL
Species Reactivity Human, Mouse, Rat
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55332, 679937, 71712
Target Species Human
Target Species Murine
Target Species Rat

DRAM Antibody, OSD00006W, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from the internal region of human DRAM
Formulation: 150μL unpurified antibody in whole serum and no preservative
Synonyms: Damage-regulated autophagy modulator, FLJ11259
Uniprot Accession: Q8N682, Q9DC58
Entrez GeneID: 55332, 679937, 71712
Gene Symbol: DRAM, DRAM1
Reconstitute with 150 ul of distilled water.