Please login to your online account to display your discounted pricing

anti-DRAM Polyclonal, Thermo Scientific™

DRAM Polyclonal Ab OSD00006W from Thermo Scientific™ specifically detects DRAM in Hu, Ms and Rt samples, and it is validated for IHC and WB

$448.05 - $448.05

Specifications

Antigen DRAM
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSD00006W

 
pierce biotechnology inc
OSD00006W
Unlabeled 150μL Each for $448.05

PIOSD00007G

 
pierce biotechnology inc
OSD00007G
Unlabeled 500μg Each for $448.05
Description & Specifications

Specifications

Antigen DRAM
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide from the internal region of human DRAM
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

DRAM Polyclonal Ab OSD00006W from Thermo Scientific™ specifically detects DRAM in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from the internal region of human DRAM
Formulation: 150μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Damage-regulated autophagy modulator, FLJ11259
Uniprot Accession: Q8N682, Q9DC58
Gene Symbol: DRAM1
Entrez GeneID: 55332, 679937, 71712
OSD00006W detects DRAM in Human, Mouse and Rat samples. OSD00006W has been successfully used in Immunohistochemistry and Western Blot procedures. OSD00006W The immunogen is A synthetic peptide from the internal region of human DRAM.