Please login to your online account to display your discounted pricing

DRAM Polyclonal Antibody, Invitrogen™


DRAM Antibody, OSD00007G, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSD00007G

Catalog No. PIOSD00007G

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Western Blot
Gene Symbols DRAM, DRAM1
Formulation 500μg purified antibody (1mg/mL) and no preservative
Storage Requirements avoiding freeze/thaw cycles.
Gene Alias Damage-regulated autophagy modulator, FLJ11259
Gene Accession No. Q8N682, Q9DC58
Species Reactivity Human, Mouse, Rat
Gene ID (Entrez) 55332, 679937, 71712
Immunogen A synthetic peptide from the internal region of human DRAM
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 500μg
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Antigen DRAM
Host Species Rabbit
Isotype IgG
Format Purified

DRAM Antibody, OSD00007G, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: A synthetic peptide from the internal region of human DRAM
Formulation: 500μg purified antibody (1mg/mL) and no preservative
Synonyms: Damage-regulated autophagy modulator, FLJ11259
Uniprot Accession: Q8N682, Q9DC58
Entrez GeneID: 55332, 679937, 71712
Gene Symbol: DRAM, DRAM1
Reconstitute with 500 ul of distilled water.