Please login to your online account to display your discounted pricing

anti-DRAM, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224758SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224758SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 55332
Gene Accession No. Q8N682
Concentration 0.5mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), cow, monkey, dog and horse,92% homologous in mouse and 85% homologous in rat and chimpanzee.
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen DRAM
Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-100 of human DRAM was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO