Please login to your online account to display your discounted pricing

anti-DRAM, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224758

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224758

  • / Each
 

Description & Specifications

Specifications

Antigen DRAM
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q8N682
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human DRAM was used as the immunogen.
Isotype IgG
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55332
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), cow, monkey, dog and horse,92% homologous in mouse and 85% homologous in rat and chimpanzee.
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat