Please login to your online account to display your discounted pricing

anti-DRAM, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-100 of human DRAM was used as the immunogen.
Antigen DRAM
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224758SS

 
novus biologicals canada
NBP224758SS
Unlabeled 0.025mg Each for $109.00

NBP224758

 
novus biologicals canada
NBP224758
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 50-100 of human DRAM was used as the immunogen.
Antigen DRAM
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), cow, monkey, dog and horse,92% homologous in mouse and 85% homologous in rat and chimpanzee.
Gene Accession No. Q8N682
Gene ID (Entrez) 55332
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL