Please login to your online account to display your discounted pricing

anti-DRAK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056695

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056695

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9262
Regulatory Status RUO
Formulation PBS
Primary or Secondary Primary
Gene Accession No. NP_004217.1
Applications Western Blotting
Quantity 0.1mg
Antigen DRAK2
Immunogen This polyclonal antibody was raised against a peptide corresponding to amino acids 351 to 365 of human DRAK2 protein.
Cross Reactivity Human
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Format Purified