Please login to your online account to display your discounted pricing

anti-DR6/TNFRSF21, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001400

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001400

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 27242
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Applications Western Blotting
Immunogen The antibody was raised against the cytoplasmic domain (amino acids 371-655) of human DR6 protein.
Format Purified
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Antigen DR6/TNFRSF21
Quantity 0.1mg
Cross Reactivity Human