Please login to your online account to display your discounted pricing

anti-DR6/TNFRSF21, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen The antibody was raised against the cytoplasmic domain (amino acids 371-655) of human DR6 protein.
Antigen DR6/TNFRSF21
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001400

 
novus biologicals canada
NB6001400
Unlabeled 0.1mg Each for $255.80

NB6001400SS

 
novus biologicals canada
NB6001400SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was raised against the cytoplasmic domain (amino acids 371-655) of human DR6 protein.
Antigen DR6/TNFRSF21
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 27242
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL