Please login to your online account to display your discounted pricing

anti-DR3/TNFRSF25, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056380

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056380

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 8718
Primary or Secondary Primary
Antigen DR3/TNFRSF25
Test Specificity In HeLa, a 50 kDa band should be observed.
Concentration 0.5mg/mL
Gene Accession No. NP_042997
Formulation PBS containing 0.2% gelatin
Cross Reactivity Human
Host Species Rabbit
Quantity 0.1mg
Regulatory Status RUO
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 398-417 of human DR3 protein.
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Purified
Applications Western Blotting