Please login to your online account to display your discounted pricing

anti-Dopamine Receptor D4/DRD4 Polyclonal, Thermo Scientific™

Dopamine Receptor D4/DRD4 Polyclonal Ab PA5-33483 from Thermo Scientific™ specifically detects Dopamine Receptor D4/DRD4 in Hu samples, and it is validated for IHC(P)

$506.05 - $506.05

Specifications

Antigen Dopamine Receptor D4/DRD4
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from C-terminus of human DRD4/Dopamine Receptor D4.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA533484

 
pierce biotechnology inc
PA533484
Unlabeled 50μg Each for $506.05

PIPA533483

 
pierce biotechnology inc
PA533483
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen Dopamine Receptor D4/DRD4
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from C-terminus of human DRD4/Dopamine Receptor D4.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

Dopamine Receptor D4/DRD4 Polyclonal Ab PA5-33483 from Thermo Scientific™ specifically detects Dopamine Receptor D4/DRD4 in Hu samples, and it is validated for IHC(P)

EDG1, a Lysophospholipid/Lysosphingolipid Receptor, binds sphingosine-1-phosphate and may control cell proliferation, survival, and suppression of apoptosis. Overexpression of EDG1 leads to increased cell-cell aggregation and overexpression of cadherins. EDG1 transcripts have been reported to be present in embryonic and adult brain, lung alveolar macrophages, vascular smooth muscle cells, endothelial cells, fibroblasts, melanocytes, and TAg-Jurkat T cells, but to be absent from breast cancer cell lines.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 17 amino acid peptide from C-terminus of human DRD4/Dopamine Receptor D4.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: DRD4, D(4) dopamine receptor, D4DR, D(2C) dopamine receptor, D4 dopamine receptor, Dopamine receptor D4, Dopamine 4 receptor, Dopamine D4 receptor
Uniprot Accession: P21917
Gene Symbol: DRD4
Entrez GeneID: 1815
PA5-34181 detects EDG1/S1P1 in Human samples. PA5-34181 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-34181 The immunogen is Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human S1PR1/EDG1..