Please login to your online account to display your discounted pricing

Dopamine Receptor D4/DRD4 Polyclonal Antibody, Invitrogen™


Dopamine Receptor D4/DRD4 Antibody, PA5-33484, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).

Manufacturer: pierce biotechnology inc  PA533484

Catalog No. PIPA533484

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Gene ID (Entrez) 1815
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Gene Symbols DRD4
Gene Alias DRD4, D(4) dopamine receptor, D4DR, D(2C) dopamine receptor, D4 dopamine receptor, Dopamine receptor D4, Dopamine 4 receptor, Dopamine D4 receptor
Species Reactivity Human
Gene Accession No. P21917
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human DRD4/Dopamine Receptor D4.
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Quantity 50μg
Format Purified
Cross Reactivity Human
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Isotype IgG
Antigen Dopamine Receptor D4/DRD4

Dopamine Receptor D4/DRD4 Antibody, PA5-33484, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry (Paraffin).

This gene encodes the D4 subtype of the dopamine receptor. The D4 subtype is a G-protein coupled receptor which inhibits adenylyl cyclase. It is a target for drugs which treat schizophrenia and Parkinson disease. Mutations in this gene have been associated with various behavioral phenotypes, including autonomic nervous system dysfunction, attention deficit/hyperactivity disorder, and the personality trait of novelty seeking. This gene contains a polymorphic number (2-10 copies) of tandem 48 nt repeats; the sequence shown contains four repeats.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human DRD4/Dopamine Receptor D4.
Formulation: 50 ug purified antibody (1mg/mL) in PBS and < 0.1% sodium azide
Synonyms: DRD4, D(4) dopamine receptor, D4DR, D(2C) dopamine receptor, D4 dopamine receptor, Dopamine receptor D4, Dopamine 4 receptor, Dopamine D4 receptor
Uniprot Accession: P21917
Entrez GeneID: 1815
Gene Symbol: DRD4
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%) Gorilla (95%) Bovine, Pig (89%) Marmoset, Dog, Horse (84%).