Please login to your online account to display your discounted pricing

anti-Dnmt3L, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227086SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227086SS

  • / Each
 

Description & Specifications

Specifications

Antigen Dnmt3L
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q9UJW3
Host Species Rabbit
Immunogen A portion of amino acids 250-300 from human Dnmt3L was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 29947
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, gibbon and chimp, 88% in pig, horse and guinea pig, 82% in Chinese hamster, 64% in rat and 53% homolgous in mouse.
Target Species Human
Target Species Primate