Please login to your online account to display your discounted pricing

anti-Dnmt3L, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227086

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227086

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 29947
Primary or Secondary Primary
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 250-300 from human Dnmt3L was used as the immunogen for this antibody.
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, gibbon and chimp, 88% in pig, horse and guinea pig, 82% in Chinese hamster, 64% in rat and 53% homolgous in mouse.
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q9UJW3
Conjugate Unlabeled
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Regulatory Status RUO
Antigen Dnmt3L
Format Purified
Applications Immunohistochemistry (Paraffin)