missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Dnmt3L, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 from human Dnmt3L was used as the immunogen for this antibody.
Antigen Dnmt3L
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227086SS Novus Biologicals Canada
NBP227086SS
0.025mg Each for $109.00
NBP227086 Novus Biologicals Canada
NBP227086
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
Dnmt3L
Purified
RUO
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, gibbon and chimp, 88% in pig, horse and guinea pig, 82% in Chinese hamster, 64% in rat and 53% homolgous in mouse.
29947
0.5mg/mL
A portion of amino acids 250-300 from human Dnmt3L was used as the immunogen for this antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin)
Q9UJW3
0.1mg antibody in 0.2mL PBS containing 0.05% BSA.