Please login to your online account to display your discounted pricing

anti-DNMT3B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100266SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100266SS

  • / Each
 

Description & Specifications

Specifications

Antigen DNMT3B
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Antisera
Gene Accession No. Q9UBC3
Host Species Rabbit
Immunogen Amino acids 4-101 of human Dnmt3b [UniProt No. Q9UBC3]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1789
Target Species Human
Target Species Murine