Please login to your online account to display your discounted pricing

anti-DNMT3B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100266

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100266

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1789
Monoclonal or Polyclonal Polyclonal
Immunogen Amino acids 4-101 of human Dnmt3b [UniProt No. Q9UBC3]
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Conjugate Unlabeled
Gene Accession No. Q9UBC3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Antiserum
Antigen DNMT3B
Quantity 0.1mL
Regulatory Status RUO
Host Species Rabbit