Please login to your online account to display your discounted pricing

anti-DNMT3B, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Amino acids 4-101 of human Dnmt3b [UniProt No. Q9UBC3]
Antigen DNMT3B
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100266SS

 
novus biologicals canada
NB100266SS
Unlabeled 0.025mL Each for $139.00

NB100266

 
novus biologicals canada
NB100266
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 4-101 of human Dnmt3b [UniProt No. Q9UBC3]
Antigen DNMT3B
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q9UBC3
Gene ID (Entrez) 1789