Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203985

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203985

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1788
Gene Accession No. NP_072046.2
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Antigen DNMT3A
Conjugate Unlabeled
Test Specificity This antibody will not cross react with Dnmt3b. Reacts with Cat, Chicken, Chimpanzee, Cow, Horse, Human and Mouse.
Primary or Secondary Primary
Quantity 0.1mg
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Host Species Rabbit
Regulatory Status RUO
Immunogen A portion of amino acids 170-220 of human Dnmt3a was used as the immunogen
Format Affinity Purified
Applications Western Blotting