Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224471SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224471SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1788
Gene Accession No. Q9Y6K1
Conjugate Unlabeled
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Test Specificity This STAT6 antibody will cross-react with mouse and rat, based on NCBI BLAST.
Immunogen A portion of amino acids 10-50 of human Dnmt3a was used as the immunogen.
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.025mg
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Antigen DNMT3A
Regulatory Status RUO