Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224471

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224471

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1788
Gene Accession No. Q9Y6K1
Formulation 100μg in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Primate
Test Specificity This STAT6 antibody will cross-react with mouse and rat, based on NCBI BLAST.
Immunogen A portion of amino acids 10-50 of human Dnmt3a was used as the immunogen.
Antigen DNMT3A
Quantity 0.1mg
Applications Western Blotting
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit