missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to areas around amino acids 125-175 and another around 175-225 of Dnmt3a.
Antigen DNMT3A
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP203987 Novus Biologicals Canada
NBP203987
0.1mg Each for $307.00
NBP203987SS Novus Biologicals Canada
NBP203987SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
DNMT3A
Purified
RUO
Cross reacts with Human. Predicted to react with Mouse.
1788
1mg/mL
This antibody was developed against synthetic peptides corresponding to areas around amino acids 125-175 and another around 175-225 of Dnmt3a.
Polyclonal
Primary
Western Blot
NP_715640
100μg in 200μL PBS containing 0.05% BSA