Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 170-220 of human Dnmt3a was used as the immunogen
Antigen DNMT3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203985

 
novus biologicals canada
NBP203985
Unlabeled 0.1mg Each for $307.00

NBP203985SS

 
novus biologicals canada
NBP203985SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 170-220 of human Dnmt3a was used as the immunogen
Antigen DNMT3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This antibody will not cross react with Dnmt3b. Reacts with Cat, Chicken, Chimpanzee, Cow, Horse, Human and Mouse.
Gene Accession No. NP_072046.2
Gene ID (Entrez) 1788
Formulation 100μg in 200μL PBS containing 0.05% BSA