Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203987SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203987SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1788
Formulation 100μg in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Regulatory Status RUO
Test Specificity Cross reacts with Human. Predicted to react with Mouse.
Gene Accession No. NP_715640
Quantity 0.025mg
Concentration 1mg/mL
Antigen DNMT3A
Applications Western Blotting
Immunogen This antibody was developed against synthetic peptides corresponding to areas around amino acids 125-175 and another around 175-225 of Dnmt3a.
Cross Reactivity Human
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Format Purified
Host Species Rabbit