Please login to your online account to display your discounted pricing

anti-DNMT3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203987

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203987

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1788
Concentration 1mg/mL
Formulation 100μg in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Cross reacts with Human. Predicted to react with Mouse.
Regulatory Status RUO
Conjugate Unlabeled
Gene Accession No. NP_715640
Format Purified
Antigen DNMT3A
Immunogen This antibody was developed against synthetic peptides corresponding to areas around amino acids 125-175 and another around 175-225 of Dnmt3a.
Applications Western Blotting
Quantity 0.1mg
Host Species Rabbit
Cross Reactivity Human